Learn More
Strem, An Ascensus Company CAS# 959395-10-9. 500mg. (2S,2'S)-(-)-[N,N'-Bis(2-pyridylmethyl]-2,2'-bipyrrolidinebis(acetonitrile)iron(II) hexafluoroantimonate Fe(S,S-PDP) White-Chen Catalyst. MFCD16038114
SDP
Supplier:  Strem, An Ascensus Company 260060500MG
Encompass
CAS# 959395-10-9. 500mg. (2S,2'S)-(-)-[N,N'-Bis(2-pyridylmethyl]-2,2'-bipyrrolidinebis(acetonitrile)iron(II) hexafluoroantimonate Fe(S,S-PDP) White-Chen Catalyst. MFCD16038114. Molecular Weight: 931.90. Molecular Formula: [C24H32FeN6]+2 (SbF6)2. Color/form: purple pwdr. Strem# 26-0060. http://www.strem.com/catalog/v/26-0060/
Catalog No. 50-201-6730