Learn More
Electronic Imaging Materials Inc 667 CryoLabel 2.50 inch x 0.38 inch, White, 10K per roll, 3 inch core
SDP
Catalog No. NC1156371
Encompass_Preferred
2.50 inch x 0.38 inch CryoLabel will handle extremely cold storage temperatures found in genetics, virology, microbiology and related fields. Conformable—works well on tight diameter test tubes. Resistant to moisture, smudging, and tearing, plus it holds up to alcohol and other chemicals. Designed for thermal transfer and durable UV inkjet printing, allowing you to color-code labels. Scratch and smear resistant when printed with T68 premium resin ribbon.
Catalog No. NC1156371 Supplier Electronic Imaging Materials Inc Supplier No. 667250038
2.50 inch x 0.38 inch CryoLabel will handle extremely cold storage temperatures found in genetics, virology, microbiology and related fields. Conformable—works well on tight diameter test tubes. Resistant to moisture, smudging, and tearing, plus it holds up to alcohol and other chemicals. Designed for thermal transfer and durable UV inkjet printing, allowing you to color-code labels. Scratch and smear resistant when printed with T68 premium resin ribbon.