Learn More
Key Scientific Products CryoCare, Organism Preservative System, White Beads, 50 Vials per Box
SDP
Catalog No. NC9997442
Encompass_Preferred
CryoCare organism preservative system lowers the cost of stock cultures by stretching each expensive culture into 15 or more which meet NCCLS standards. Vial contains ceramic beads in broth base. Includes 50 vials per box with white beads.
Catalog No. NC9997442 Supplier Key Scientific Products Supplier No. KS70WH50
CryoCare organism preservative system lowers the cost of stock cultures by stretching each expensive culture into 15 or more which meet NCCLS standards. Vial contains ceramic beads in broth base. Includes 50 vials per box with white beads.