Learn More
NEXPRING HEALTH White Daisy Goblets with Visotubes, 5 pack
Catalog No. NC2173906
Encompass
Daisy Goblets with Visotubes are used for freezing and storage of biological materials in straws. Arrangement using color coding of straws prior to storage in liquid nitrogen. Daisy goblet (perforated goblet, white cover and 12 visotubes) Ø: 65mm (67mm with cover) Goblet height: 117mm Total height of daisy goblet (with cover): 135mm Contains: 5 daisy goblets, 11 triangular colored visotubes (120mm), 1 round clear visotube diameter 17.5mm (133mm).
Catalog No. NC2173906 Supplier NEXPRING HEALTH Supplier No. 015152
Daisy Goblets with Visotubes are used for freezing and storage of biological materials in straws. Arrangement using color coding of straws prior to storage in liquid nitrogen. Daisy goblet (perforated goblet, white cover and 12 visotubes) Ø: 65mm (67mm with cover) Goblet height: 117mm Total height of daisy goblet (with cover): 135mm Contains: 5 daisy goblets, 11 triangular colored visotubes (120mm), 1 round clear visotube diameter 17.5mm (133mm).