Learn More
Thomas Scientific White Dots for 0.5 - 1.5mL Tubes, 9.5mm, 1000/Roll
SDP
Catalog No. 501460190
Encompass
PolarSafe Labels Rolls Argos PolarSafe Dots and Labels are made from chemically inert polyester with a unique adhesive formulated to adhere to laboratory glass and plasticware. These unique labels will not only withstand conventional freezing (-80°C) but can also be used for most cryogenic storage applications (vapor phase nitrogen storage at -196°C). No need for separate labeling solutions for your cryo-preservation units and your ultra low freezers. The labels can even be used in incubators. Argos PolarSafe Labels do not require self lamination to be effective. The sleeker labels mean your tubes will still fit most storage systems without the label sticking or being torn off. The wide range of sizes allow you to label your whole experiment permanently, from micro-centrifuge tube to cryovial; microtiter plate to storage boxes. Argos PolarSafe Label Dispenser Introducing the newest product addition to the PolarSafe family of products, the Argos label dispenser can accommodate u
Catalog No. 50-146-0190 Supplier Thomas Scientific Supplier No. 1213N00
PolarSafe Labels Rolls Argos PolarSafe Dots and Labels are made from chemically inert polyester with a unique adhesive formulated to adhere to laboratory glass and plasticware. These unique labels will not only withstand conventional freezing (-80°C) but can also be used for most cryogenic storage applications (vapor phase nitrogen storage at -196°C). No need for separate labeling solutions for your cryo-preservation units and your ultra low freezers. The labels can even be used in incubators. Argos PolarSafe Labels do not require self lamination to be effective. The sleeker labels mean your tubes will still fit most storage systems without the label sticking or being torn off. The wide range of sizes allow you to label your whole experiment permanently, from micro-centrifuge tube to cryovial; microtiter plate to storage boxes. Argos PolarSafe Label Dispenser Introducing the newest product addition to the PolarSafe family of products, the Argos label dispenser can accommodate u