Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Spilfyter™ White DRC Wiper

Catalog No. 17000274 Shop All NPS Corporation Products
Change view
Click to view available options
Color:
White
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
17-000-274 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-000-274 Supplier Spilfyter™ Supplier No. D12110
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Spilfyter White DRC Wiper is a tough general purpose wipe with a cloth like feel.

  • Constructed from cellulose fibers with a binder for additional strength
  • Cloth-like disposable wipers
  • Superior oil and grease clean up
  • Excellent wet strength and wipe dry properties make them ideal for surface and parts cleaning
  • Highly absorbent and durable to tackle everyday tasks

Specifications

Material Double Re-crepe Cellulose
Color White
Container Type Corrugate Box
Disposable Single-use
Dimensions (L x W) 9.25 x 12.5 in. (23.5 x 31.75 cm)
For Use With (Application) All-Purpose Wipe
Packaging Quantity 110 Wipes/Dispenser, 12 Boxes/Cs.
Sterility Non-sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.