Learn More
TrippNT HPLC 105 Column HDPE Column Cabinet with Lockable Hinged Door
SDP
Catalog No. NC1155493 Shop All TrippNT Products
Encompass_Preferred
TrippNT's HPLC 105 Column HDPE Column Cabinet with Lockable Hinged Door protects, stores and organizes columns. This storage cabinet allows for inventory to be easily seen and features a hasp lock. Is wall mounted using keyholes and included mounting hardware.
Catalog No. NC1155493 Supplier TrippNT Supplier No. 51409
TrippNT's HPLC 105 Column HDPE Column Cabinet with Lockable Hinged Door protects, stores and organizes columns. This storage cabinet allows for inventory to be easily seen and features a hasp lock. Is wall mounted using keyholes and included mounting hardware.