Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Qorpak™ White HDPE Wide Mouth Round Bottles without Cap

Catalog No. 02993502
Change view
Click to view available options
Capacity (Metric):
2000 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
02-993-502 2000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 02-993-502 Supplier Qorpak™ Supplier No. 241385

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ideal for general lab use with solids and powders. Qorpak™ White HDPE Wide Mouth Round Bottles without Cap provide moisture barrier and are impact resistant.

  • Ideal for high-speed filling, demanding pharmaceutical specifications and rough handling of interstate shipping

Specifications

Capacity (Metric) 2000 mL
Color White
Material HDPE
Neck Finish 110-400
Capacity (English) 68 oz.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.