Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Hose Clamp, Acetal Copolymer, White, 0.569"" ID x 0.650"" ID; 100/PK
Catalog No. NC9719040
Encompass
These reusable white acetal copolymer clamps will not rust or corrode. They are recommended for continuous use from ambient temperature to 200F or 93C. Clamps are nonconducting and stable in a wide variety of chemicals.Once set, clamps stay put. To use, simply position the clamp on a hose or tubing bundle and then press the jaws together with a pair of pliers.
Catalog No. NC9719040 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0683210
These reusable white acetal copolymer clamps will not rust or corrode. They are recommended for continuous use from ambient temperature to 200F or 93C. Clamps are nonconducting and stable in a wide variety of chemicals.Once set, clamps stay put. To use, simply position the clamp on a hose or tubing bundle and then press the jaws together with a pair of pliers.