Learn More
Andwin Scientific WHITE LASER CRYO-BABIES SMAL
SDP
Catalog No. 50821019
Encompass_Preferred
The Cryo-Tags white labels measure 0.94" x 0.50". This product features compatibility with laser printers as the labels are heat-resistant and have a stay-flat design. The labels can withstand temperatures from -196C to 100C and can edure boiling water baths, oven temperatures, and autoclaving and freezer temperatures. The Cryo-Tags can also combat most organic solvents and caustic agents. Compatible with most surfaces, like plastics, glass, and metals without cracking, peeling, or degrading.
Catalog No. 50-821-019 Supplier Andwin Scientific Supplier No. 91872380
The Cryo-Tags white labels measure 0.94" x 0.50". This product features compatibility with laser printers as the labels are heat-resistant and have a stay-flat design. The labels can withstand temperatures from -196C to 100C and can edure boiling water baths, oven temperatures, and autoclaving and freezer temperatures. The Cryo-Tags can also combat most organic solvents and caustic agents. Compatible with most surfaces, like plastics, glass, and metals without cracking, peeling, or degrading.