Learn More
Andwin Scientific WHITE LASER TOUGH-SPOTS LARG
SDP
Supplier:  Andwin Scientific 91852000
Encompass_Preferred
The white Tough-Spots labels are specially designed to be used for 0.5 mL tubes. The labels are resistant against chemicals and temperatures from -196C to 80C. This product can be put on several surfaces like most plastics, glass, vinyl, and metals. Any marking instrument can also be utilized to write on these labels. This product has a diameter of 1/2".
Catalog No. 50-820-993