Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Qorpak™ White Metal Pulp/Polyethylene Lined Caps

Catalog No. 02993390
Change view
Click to view available options
Neck Finish:
100-400
38-400
43-400
48-400
58-400
63-400
70-400
89-400
No. per Case:
1400 Cs.
144 Cs.
288 Cs.
500 Cs.
576 Cs.
Packaging:
Bagged
Bulk
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Neck Finish No. per Case Packaging
02-993-390 48-400 576 Cs. Bagged
02-993-383 58-400 288 Cs. Bagged
02-993-380 70-400 144 Cs. Bagged
02-991-331 100-400 500 Cs. Bulk
02-993-379 63-400 288 Cs. Bagged
02-993-365 38-400 576 Cs. Bagged
02-991-090 70-400 1400 Cs. Bulk
02-993-419 89-400 144 Cs. Bagged
02-993-389 43-400 576 Cs. Bagged
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 02-993-390 Supplier Qorpak™ Supplier No. CAP06659
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Exhibit tolerance to widest range of temperatures

  • Resistant to fracture from impact
  • Offered in 400 GPI thread finish
  • For general-purpose
  • Ideal for powders, non-acidic food products and consumer products

Specifications

Color White
Material Metal
No. per Case 576 Cs.
No. per Pack 12/Pk.
Packaging Bagged
Sterility Non-sterile
Liner Material Pulp/Polyethylene
Neck Finish 48-400
For Use With (Equipment) Wide mouth bottle
Autoclavable Not Autoclavable
Disposable Reusable
Product Type Threaded Screw Cap
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.