Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ White Paper Ream for Science Kits
SCI_ED

Catalog No. S05176 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 Sheets
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S05176 500 Sheets
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S05176
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Consumables for use with multiple projects and in science kits.

  • 8.5 x 11
  • 500 sheets per pack and print ready
  • White paper with matte finish
  • Acid-free to prevent crumbling or yellowing

Specifications

Product Type Paper
Color White
For Use With (Application) For multiple projects or in a science kit
Quantity 500 Sheets
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.