Learn More
Valutek™ White Polyester Microfiber Filament Flat Mop
Catalog No. 191502129
Change view
Click to view available options
Dimensions:
4.5 x 16 in.
Catalog No. Dimensions
19-150-2129 4.5 x 16 in.
1 options
Catalog No. 19-150-2129 Supplier Valutek™ Supplier No. VTCRMOPF4516
Add to Cart
Add to Cart
Head is manufactured with 1/4 in. knitted tube. This mop head is made of 100% white filament polyester looped to form edgeless tubular strands to absorb and encapsulate liquids for maximum sorptive capacity.
  • This mop head has a flat design that slides easily on walls or floors
  • Superb absorbency
  • Ultra-low particulate and fiber generation
  • Flat design slides easily and lowers time and labor consumption
  • ESD safe
  • The knitted fabric is autoclavable and has extremely low particle and fiber generation which ensures excellent cleaning results
  • Laundered and packaged in a cleanroom
  • All Valutek™ mop heads are tested and manufactured in ISO compliant facilities under Valutek™ inspection and strict process control to ensure Valutek™ quality standards and product specifications
  • This product is recommended for use in a cleanroom Class 1-10 (ISO 3-4) critical environment
  • This mop head’s unique flat design provides for smooth sliding and makes it ideal for wiping perforated floor tiles in micro-electronics applications
  • The mop head fits standard wall mop handles
Specifications
Dimensions 4.5 x 16 in.
Sterility Non-sterile
Material Polyester