Learn More
AIR CONTROL INC White polyproLABS Cabinet
SDP
Supplier:  AIR CONTROL INC AC2053030
Encompass_Preferred
Air Control’s polyproLABS line of corrosion resistant polypropylene (polypro) casework and clean storage systems are the best available lab furniture on the market for caustic, critical processing environments. Entire TRACE METALS ANALYSIS lab o ra to ries may be specified as having NO EXPOSED METALS, in any of our equipment and furniture. All hardware including door hinges, latches, drawer suspension sys tems, handles, and plumbing accessories are made from polypropylene. Optioal (not included) worksurfaces are available and unique to our line of case work. All workmanship and components are designed, built, and 3rd party tested in the USA in accordance with the SEFA 8 standard of construction for the laboratory casework industry. MADE IN USA. Other sizes available.
Catalog No. NC3311192