Learn More
VGuard™ White Polypropylene Arm Sleeve
Catalog No. 17005959
Encompass
Encompass_Preferred
This item is not returnable. View return policy
Catalog No. 17-005-959 Supplier VGuard™ Supplier No. B16M10
This item is not returnable. View return policy
Breathable, comfortable, and lightweight arm protection.
  • Disposable
  • Durable Spunbond Polypropylene for all-day wear
  • Keeps skin and hair from contaminating product
  • Protects against dust, dirt, grime, and light fluid spray
Specifications
Product Type General Purpose Sleeve
Material Spunbond Polypropylene
Color White
Sterility Non-sterile
Disposable Single-use
Closure Type Elastic Ends
Certifications/Compliance 21 CFR, Parts 170-199
Length (English) 18 in.
Length (Metric) 457.2 mm