Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wheaton™ White Polypropylene Screw Caps
SDP

Catalog No. 12141619 Shop All DWK Life Sciences Products
Change view
Click to view available options
Color:
White
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
12-141-619 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12-141-619 Supplier Wheaton™ Supplier No. 300203
Add to Cart
Add to Cart

Suitable for most applications that call for screw neck bottles. Wheaton™ White Polypropylene Screw Caps are manufactured from sturdy polypropylene plastic.

  • For thread size 22mm
  • Ideal for storage and containment of volatile solvents
  • Excellent closing option for screw neck bottles and vials
TRUSTED_SUSTAINABILITY

Specifications

Color White
Material Polypropylene
For Use With (Application) For screw thread diagnostic vials
Thread Style 22 mm
Closure Type Cap
Quantity 1000/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.