Learn More
Fisherbrand™ White Polypropylene Caps
SDP
Catalog No. FB02912068 Shop All Fisherbrand Products
Change view
Click to view available options
Neck Finish:
15-425
18-400
20-400
22-400
24-400
28-400
33-400
38-400
43-400
45-400
48-400
53-400
58-400
63-400
70-400
83-400
89-400
Liner Material:
PTFE/Polyethylene Foam
Polyethylene Foam
Pulp/Vinyl
Quantity:
100/Cs.
1300/Cs.
144/Cs.
2100/Cs.
288/Cs.
370/Cs.
500/Cs.
23 product options available for selection
Product selection table with 23 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Neck Finish Liner Material Quantity
FB02912068 22-400 PTFE/Polyethylene Foam 500/Cs.
FB02911836A 33-400 PTFE/Polyethylene Foam 2100/Cs.
FB02911844A 53-400 Pulp/Vinyl 1300/Cs.
FB02912066 18-400 PTFE/Polyethylene Foam 100/Cs.
FB02912065 15-425 PTFE/Polyethylene Foam 500/Cs.
FB02912067 20-400 PTFE/Polyethylene Foam 100/Cs.
FB02912069 24-400 PTFE/Polyethylene Foam 100/Cs.
FB02912070 28-400 PTFE/Polyethylene Foam 100/Cs.
FB02912071 33-400 PTFE/Polyethylene Foam 100/Cs.
FB02912072 38-400 PTFE/Polyethylene Foam 500/Cs.
FB02912073 43-400 PTFE/Polyethylene Foam 500/Cs.
FB02912074 45-400 PTFE/Polyethylene Foam 500/Cs.
FB02912076 53-400 PTFE/Polyethylene Foam 100/Cs.
FB02912075 48-400 PTFE/Polyethylene Foam 100/Cs.
FB02912077 58-400 PTFE/Polyethylene Foam 100/Cs.
FB02912078 70-400 PTFE/Polyethylene Foam 144/Cs.
FB02912079 83-400 PTFE/Polyethylene Foam 144/Cs.
FB02912080 89-400 PTFE/Polyethylene Foam 144/Cs.
FB02912085 63-400 PTFE/Polyethylene Foam 288/Cs.
FB02912093A 53-400 Polyethylene Foam 1300/Cs.
FB02912099 53-400 Polyethylene Foam 500/Cs.
FB02912094A 89-400 Polyethylene Foam 370/Cs.
FB02912292A 53-400 Pulp/Vinyl 288/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 23 options available.
23 options
Catalog No. FB02912068
Only null left
Add to Cart
Add to Cart
White polypropylene screw caps provides clear surface for writing on cap
  • Foamed polyethylene liner provides tight seal and chemical resistance for acids, alkalis, solvents, alcohols, oils, household cosmetics and aqueous products
  • PTFE faced foamed polyethylene liner offers chemical resistance excellent for environmental samples, pharmaceuticals and diagnostic reagents
  • Poly-vinyl liner provides an excellent moisture barrier good for mild acids, alkalis, solvents, alcohols, oils and aqueous products
TRUSTED_SUSTAINABILITY
Specifications
Color White
Material Polypropylene
Quantity 500/Cs.
Liner Material PTFE/Polyethylene Foam
Neck Finish 22-400
Product Type Lined Cap