Learn More
Valutek™ Spunbond Polypropylene Lab Coats
Catalog No. 191502089
This item is not returnable. View return policy
Catalog No. 19-150-2089 Supplier Valutek™ Supplier No. VTLBCT3X
Add to Cart
Add to Cart
This item is not returnable. View return policy
Disposable lab coats without pockets are manufactured with strong, lightweight, breathable, non-linting fabric.
  • Lab coats are knee length with 5 snap closures, elastic cuffs, and a single ply shirt collar design
  • Without pockets
  • Tested and manufactured in ISO compliant facilities to ensure quality standard and product specification compliance
  • Recommended for use in a cleanroom Class 10,000+ (ISO 7+) critical environments
  • Commonly used in electronic, medical device, and pharmaceutical industries where operator garment contamination is a concern
  • Highly suitable for a wide variety of macro contamination level applications, such as basic manufacturing, MDM and injection molding. 
Specifications
Product Type Controlled Environments Lab Coat
Material Polypropylene
Color White
Cuff Style Elastic
Disposable Single-use
Sterility Non-sterile
Collar Type Shirt Collar Design
Cleanroom Class Class 10,000 (ISO 7)