Learn More
Qorpak™ White Polypropylene Unlined Cap
Catalog No. 5001167
Change view
Click to view available options
Includes:
Standard Ribbing
Catalog No. Includes
5001167 Standard Ribbing
1 options
Catalog No. 5001167 Supplier Qorpak™ Supplier No. CPD03812
Add to Cart
Edge
Add to Cart
Suitable for dry products. Qorpak™ White Polypropylene Unlined Cap are known for good impact strength and pliability.
  • Autoclavable
  • Available in ribbed and fine ribbed style
Specifications
Color White
Includes Standard Ribbing
Material Polypropylene
Sterility Non-sterile
Liner Material Unlined
Neck Finish 110-400
Autoclavable Not Autoclavable
Disposable Reusable
Product Type Ribbed Cap