Learn More
Bellco Glass, Inc. White PTFE Septa Disk, 8mm
SDP
Catalog No. 501945391
Encompass_Preferred
Manufactured from red ptfe silicone. Ideal for screw thread vials. MADE IN THE USA. Autoclavable Size: 8mm Septa thickness: 0.010"
Catalog No. 50-194-5391 Supplier Bellco Glass, Inc. Supplier No. 207001100
Manufactured from red ptfe silicone. Ideal for screw thread vials. MADE IN THE USA. Autoclavable Size: 8mm Septa thickness: 0.010"