Learn More
MSC White Rodgers Generator, 750 mV, 36 in Leads, 1 ea
Catalog No. 5000504673
Encompass
This power pile generator provides power for 750 millivolt self-generation control systems. It is designed for replacement of similar screw-in type generators currently in the field or for use with a type E31-12 pilot burner.
  • Provides power for 750 millivolt self-generation control systems
  • Designed for replacement of similar screw-in type generators or use with type E31-12 pilot burner
  • Features 36-inch fiberglass leads with spade terminals
Catalog No. 50-005-04673 Supplier MSC Supplier No. 05287263
This power pile generator provides power for 750 millivolt self-generation control systems. It is designed for replacement of similar screw-in type generators currently in the field or for use with a type E31-12 pilot burner.
  • Provides power for 750 millivolt self-generation control systems
  • Designed for replacement of similar screw-in type generators or use with type E31-12 pilot burner
  • Features 36-inch fiberglass leads with spade terminals