This universal hot surface ignition control is designed for gas-fired applications. Its microprocessor-based design ensures maximum compatibility with existing systems, offering programmable timing via program keys and a diagnostic LED for quick troubleshooting. It manages the ignitor and gas valve, featuring a 100% shut-off for reliable and safe operation if ignition fails.
- Universal replacement for many OEM and competitive models.
- Programmable timing for ignition trial, retries, prepurge, and igniter warm-up.
- LED indicator for system diagnostics and status.
- Compatible with direct or indirect flame sensing systems.
- 100% shut-off ensures safety if burner fails to ignite.
- Approved for natural, manufactured, mixed, liquid petroleum, and LP gas-air mixtures.
- Operates with 18 to 30 VAC input voltage.
- Designed for surface mount or 4" x 4" junction box installation.