Learn More
MSC White-Rodgers Hydronic Strap-On Control, 10 F Differential, 100-240, 1 ea
Catalog No. 5000504936
Encompass
This hydronic zone control is designed for strap-on applications, providing temperature regulation for heating and cooling systems. It features a precise differential and a broad operating temperature range, making it suitable for various tube sizes.
  • Designed for high-speed applications
  • Suitable for any tube size with strap-on mounting
  • Features a 10°F (5.5°C) differential
  • Operates within a temperature range of 100°F to 240°F
  • Compatible with 24V AC, 120V AC, and 240V AC systems
Catalog No. 50-005-04936 Supplier MSC Supplier No. 05423397
This hydronic zone control is designed for strap-on applications, providing temperature regulation for heating and cooling systems. It features a precise differential and a broad operating temperature range, making it suitable for various tube sizes.
  • Designed for high-speed applications
  • Suitable for any tube size with strap-on mounting
  • Features a 10°F (5.5°C) differential
  • Operates within a temperature range of 100°F to 240°F
  • Compatible with 24V AC, 120V AC, and 240V AC systems