Learn More
MSC White-Rodgers Motor Assembly, Hydronic Zone Valve, 2-Wire, 24V, 1 ea
Catalog No. 5000505860
Encompass
The F19-0104 is a motor assembly designed as a replacement part for White-Rodgers 1300 Series Hydronic Zone Valves. It is specifically intended for use with 2-wire zone valves, including models 1361-102, -103, and -104, when the valve mechanism fails to operate correctly.
  • Designed for 1300 Series Hydronic Zone Valves
  • Compatible with 2-wire zone valves
  • Serves as a replacement when valve fails to open or close
  • Operates on 24V
  • Water seal replacement kit not included
Catalog No. 50-005-05860 Supplier MSC Supplier No. 07427719
The F19-0104 is a motor assembly designed as a replacement part for White-Rodgers 1300 Series Hydronic Zone Valves. It is specifically intended for use with 2-wire zone valves, including models 1361-102, -103, and -104, when the valve mechanism fails to operate correctly.
  • Designed for 1300 Series Hydronic Zone Valves
  • Compatible with 2-wire zone valves
  • Serves as a replacement when valve fails to open or close
  • Operates on 24V
  • Water seal replacement kit not included