Learn More
MSC White-Rodgers Snap Disc Control, 140-180F Cut-in/Close-on-Rise, 120/240 VAC, 1 ea
Catalog No. 5000504246
Encompass
The White-Rodgers snap disc control is designed for fan control applications, featuring a cut-in temperature range of 140 to 180°F and a close-on-rise switch action.
  • Automatic reset for adjustable settings
  • Close on rise switch action
  • Operates at 120/240 VAC
  • Current at full load: 10A at 240VAC; 14A at 120VAC
  • Locked rotor amperage: 72A at 120VAC, 60A at 240VAC
  • Resistive rating: 25A at 120VAC, 25A at 240VAC
  • Cut-in temperature: 140 to 180°F
Catalog No. 50-005-04246 Supplier MSC Supplier No. 05423231
The White-Rodgers snap disc control is designed for fan control applications, featuring a cut-in temperature range of 140 to 180°F and a close-on-rise switch action.
  • Automatic reset for adjustable settings
  • Close on rise switch action
  • Operates at 120/240 VAC
  • Current at full load: 10A at 240VAC; 14A at 120VAC
  • Locked rotor amperage: 72A at 120VAC, 60A at 240VAC
  • Resistive rating: 25A at 120VAC, 25A at 240VAC
  • Cut-in temperature: 140 to 180°F