Learn More
MSC White-Rodgers Snap Disc Control, 175-215F, Open on Rise, 1 ea
Catalog No. 5000503537
Encompass
This snap disc adjustable temperature fan limit control features an automatic reset and an open on rise switch action, providing reliable temperature regulation for various heating equipment and appliances.
  • Adjustable temperature setting
  • Automatic reset functionality
  • Open on rise switch action
  • Equipped with 1/4 inch quick connect terminals
  • Reduces inventory needs by covering a wide range of applications
  • Replaces most fixed disc thermostats
Catalog No. 50-005-03537 Supplier MSC Supplier No. 05423272
This snap disc adjustable temperature fan limit control features an automatic reset and an open on rise switch action, providing reliable temperature regulation for various heating equipment and appliances.
  • Adjustable temperature setting
  • Automatic reset functionality
  • Open on rise switch action
  • Equipped with 1/4 inch quick connect terminals
  • Reduces inventory needs by covering a wide range of applications
  • Replaces most fixed disc thermostats