Learn More
MSC White-Rodgers Snap Disc Control, Open on Rise, 210-250F, 1 ea
Catalog No. 5000504873
Encompass
This White-Rodgers snap disc control features an adjustable temperature set point range of 210°F to 250°F with 'Open on Rise' switch action and automatic reset. It is designed for temperature limit sensing applications in heating equipment and various appliances.
  • Equipped with 1/4 in quick connect terminals
  • Automatic reset functionality
  • Suitable for 120/240 V operation
  • Resistive rating of 25A at 120 VAC and 240 VAC
  • Replaces most fixed disc thermostats in heating equipment and various appliances
  • Features a differential of 40°F (22°C)
Catalog No. 50-005-04873 Supplier MSC Supplier No. 05423280
This White-Rodgers snap disc control features an adjustable temperature set point range of 210°F to 250°F with 'Open on Rise' switch action and automatic reset. It is designed for temperature limit sensing applications in heating equipment and various appliances.
  • Equipped with 1/4 in quick connect terminals
  • Automatic reset functionality
  • Suitable for 120/240 V operation
  • Resistive rating of 25A at 120 VAC and 240 VAC
  • Replaces most fixed disc thermostats in heating equipment and various appliances
  • Features a differential of 40°F (22°C)