Learn More
MSC White-Rodgers Thermostat, Digital Nonprogrammable, 1 Heat, 1 Cool, 45-90°F
Catalog No. 5000507074
Encompass
This 70 Series economy digital thermostat is designed for single-stage systems. It is non-programmable, features a smart design to cover wall marks from older thermostats, and has a lower profile.
  • Large, easy-to-read LCD
  • Lighted display
  • Millivolt compatible
  • Single stage heat pump (B&O terminals)
  • Supports 1 heat and 1 cool setting
  • Temperature range of 45 to 90°F
  • Operates on 20 to 30 Volts
Catalog No. 50-005-07074 Supplier MSC Supplier No. 74559782
This 70 Series economy digital thermostat is designed for single-stage systems. It is non-programmable, features a smart design to cover wall marks from older thermostats, and has a lower profile.
  • Large, easy-to-read LCD
  • Lighted display
  • Millivolt compatible
  • Single stage heat pump (B&O terminals)
  • Supports 1 heat and 1 cool setting
  • Temperature range of 45 to 90°F
  • Operates on 20 to 30 Volts