Learn More
MSC White-Rodgers Thermostat, Programmable Touchscreen, 1 Heat, 1 Cool, 45-99°F
Catalog No. 5000507350
Encompass
This programmable touchscreen thermostat is designed for precise control of various heating and cooling systems. It offers a wide temperature operating range, flexible programming choices, and energy-saving features to enhance comfort and efficiency in residential or commercial settings.
  • Programmable touchscreen interface
  • Supports 1 heat, 1 cool systems
  • Temperature range: 45 to 99°F
  • Operating ambient: 32°F to 105°F
  • Flexible scheduling: non-programmable, 5/1/1 day, or 7 day
  • Energy management recovery
  • Adjustable cycle rates
  • Compressor lockout protection
  • Continuous display backlight
  • Fahrenheit/Celsius display selectable
  • Automatic daylight saving time adjustment
  • Cool savings feature
  • Temperature limits and keypad lockout
  • Change filter reminder
  • Flexible fan and system settings
Catalog No. 50-005-07350 Supplier MSC Supplier No. 78365988
This programmable touchscreen thermostat is designed for precise control of various heating and cooling systems. It offers a wide temperature operating range, flexible programming choices, and energy-saving features to enhance comfort and efficiency in residential or commercial settings.
  • Programmable touchscreen interface
  • Supports 1 heat, 1 cool systems
  • Temperature range: 45 to 99°F
  • Operating ambient: 32°F to 105°F
  • Flexible scheduling: non-programmable, 5/1/1 day, or 7 day
  • Energy management recovery
  • Adjustable cycle rates
  • Compressor lockout protection
  • Continuous display backlight
  • Fahrenheit/Celsius display selectable
  • Automatic daylight saving time adjustment
  • Cool savings feature
  • Temperature limits and keypad lockout
  • Change filter reminder
  • Flexible fan and system settings