Learn More
Uline White Round Wide-Mouth Plastic Jars Bulk Pack - 10 oz, White Cap, 140/case
Catalog No. NC3847856
Encompass
Excellent for cosmetics, chemicals or nuts and bolts.Wide mouth, no inside lip for easy filling and pouring.Chemical-resistant polypropylene.FDA compliant.Caps included.
Catalog No. NC3847856 Supplier Uline Supplier No. S17039BW
Excellent for cosmetics, chemicals or nuts and bolts.Wide mouth, no inside lip for easy filling and pouring.Chemical-resistant polypropylene.FDA compliant.Caps included.