Learn More
Bioworld White's Medium, 50 L
Catalog No. 50254814
Encompass_Preferred
White's MediumContains Macronutrients and MicronutrientsSucrose, Cytokines, Vitamins, and Auxins should be added as needed Provided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture testedUsually ships within 24 hours This product is sold for laboratory research use only
Catalog No. 50-254-814 Supplier Bioworld Supplier No. 306301244
White's MediumContains Macronutrients and MicronutrientsSucrose, Cytokines, Vitamins, and Auxins should be added as needed Provided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture testedUsually ships within 24 hours This product is sold for laboratory research use only