Learn More
Research Products International Corp White's Medium, 9.6 Grams of Powder, Makes 10 Liters of Solution
SDP
Catalog No. 50213738
Encompass_Preferred
White P.R., The cultivation of Animal and Plant Cells, Ronald Press, New York (1963).
  • Concentration: 963.3 mg/L
Catalog No. 50-213-738 Supplier Research Products International Corp Supplier No. W1020010.0
White P.R., The cultivation of Animal and Plant Cells, Ronald Press, New York (1963).
  • Concentration: 963.3 mg/L