Learn More
MSC CRC 1004789 8 oz Tube White RTV Silicone Gasket Sealant -76 to 356°F Operating Temp, 24 hr Full Cure Time
Catalog No. 501305726
Encompass
CRC 1004789 8 oz Tube White RTV Silicone Gasket Sealant -76 to 356°F Operating Temp, 24 hr Full Cure Time
Catalog No. 50-130-5726 Supplier MSC Supplier No. 01292267
CRC 1004789 8 oz Tube White RTV Silicone Gasket Sealant -76 to 356°F Operating Temp, 24 hr Full Cure Time