Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich White spirit

Catalog No. 8646025L Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 L
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
86-460-25L 2.5 L
86-460-1L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 86-460-25L Supplier Sigma Aldrich Supplier No. 864602.5L
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • White spirit is generally used:
  • As a solvent for the extraction of oil-bituminous rock organic components from mineral
  • As a component in the mixture of organic liquids to clean the coal samples by float/sink gravity separations.white spirit with ~17% aromatics basis can be used as a solvent in the synthesis of alkyd resin from camelina oil.download our flavors and fragrances catalog to view our entire product line.subscribe to our newsletter to keep up to date on our latest flavors and fragrances offerings

Specifications

Density 0.78 g/mL (at 20°C)
Boiling Point 150°C to 200°C
Refractive Index n20/D 1.437
MDL Number MFCD00132767
Quantity 2.5 L
Percent Purity ∼17%aromatics basis
Biological Activity Central Nervous System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.