Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sarstedt Inc WHITE STOPPERS 13MM 1000/PK

Catalog No. NC0398117 Shop All Sarstedt Inc. Products
Encompass

Stopper for Tube 13mm White (Encompass)

Catalog No. NC0398117 Supplier Sarstedt Inc Supplier No. 65.806.312
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Stopper for Tube 13mm White (Encompass)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.