Learn More
Thomas Scientific White Strips for 1.5 - 2.0mL tubes, 33 x 13mm, 85/Sheet
SDP
Catalog No. 501460196
Encompass_Preferred
PolarSafe Labels For Laser Printers Argos PolarSafe Dots and Labels are made from chemically inert polyester with a unique adhesive formulated to adhere to laboratory glass and plastic ware. These unique labels will not only withstand conventional freezing (-80°C) but can also be used for most cryogenic storage applications (vapor phase nitrogen storage at -196°C). No need for separate labeling solutions for your cryo-preservation units and your ultra low freezers. The labels can even be used in incubators. Argos Micryo Labels do not require self lamination to be effective. The sleeker labels mean your tubes will still fit most storage systems without the label sticking or being torn off. The wide range of sizes allow you to label your whole experiment permanently, from micro-centrifuge tube to cryovial; microtiter plate to storage boxes.
Catalog No. 50-146-0196 Supplier Thomas Scientific Supplier No. 1213N04
PolarSafe Labels For Laser Printers Argos PolarSafe Dots and Labels are made from chemically inert polyester with a unique adhesive formulated to adhere to laboratory glass and plastic ware. These unique labels will not only withstand conventional freezing (-80°C) but can also be used for most cryogenic storage applications (vapor phase nitrogen storage at -196°C). No need for separate labeling solutions for your cryo-preservation units and your ultra low freezers. The labels can even be used in incubators. Argos Micryo Labels do not require self lamination to be effective. The sleeker labels mean your tubes will still fit most storage systems without the label sticking or being torn off. The wide range of sizes allow you to label your whole experiment permanently, from micro-centrifuge tube to cryovial; microtiter plate to storage boxes.