Learn More
MSC White Super Polish Pad 4100N, 3M, 19 in, 5 Pads
Catalog No. 5000506817
Encompass
The 3M White Super Polish Pad 4100N is a durable floor pad designed for light cleaning, buffing, and polishing. It effectively removes scuffs, soil, and black heel marks without dulling floor finishes, making it ideal for routine maintenance and enhancing floor appearance. It can be used for spray cleaning or dry buffing soft finishes, including polishing soft waxes on wood floors.
  • Made of polyester fibers in an open textured non-woven construction
  • Can be washed and reused for extended life
  • For use with low speed rotary or automatic floor machines (175–600 rpm)
  • Effectively removes scuffs, soil, and black heel marks
  • Ideal for polishing soft waxes on wood floors
Catalog No. 50-005-06817 Supplier MSC Supplier No. 73448268
The 3M White Super Polish Pad 4100N is a durable floor pad designed for light cleaning, buffing, and polishing. It effectively removes scuffs, soil, and black heel marks without dulling floor finishes, making it ideal for routine maintenance and enhancing floor appearance. It can be used for spray cleaning or dry buffing soft finishes, including polishing soft waxes on wood floors.
  • Made of polyester fibers in an open textured non-woven construction
  • Can be washed and reused for extended life
  • For use with low speed rotary or automatic floor machines (175–600 rpm)
  • Effectively removes scuffs, soil, and black heel marks
  • Ideal for polishing soft waxes on wood floors