Learn More
Andwin Scientific WHITE TIPS/53509-070 1000/PK
SDP
Catalog No. NC9536100
Encompass_Preferred
Tip, Ultra-Micro; ; Compatible with most micropipettors; Volume: 0.1 to 10uL; 2 and 10uL graduations; Manufactured from polypropylene; Nonsterile; Clear; 1000/PK
Catalog No. NC9536100 Supplier Andwin Scientific Supplier No. 53509070
Tip, Ultra-Micro; ; Compatible with most micropipettors; Volume: 0.1 to 10uL; 2 and 10uL graduations; Manufactured from polypropylene; Nonsterile; Clear; 1000/PK