Learn More
Electron Microscopy Sciences White Translucent Cutting Mat, 12x18"
SDP
Catalog No. 5033316
Encompass_Preferred
Cutting Mats will protect all table-top surfaces from knife cuts. Its smooth surface permits straight, even cutting and the cut mark heals instantly. Each mat is printed with both 1/2" and 1" grid patterns to aid in measuring and aligning work. Available in translucent white, which allows for cutting and trimming on a light box surface.
Catalog No. 50-333-16 Supplier Electron Microscopy Sciences Supplier No. 7762118
Cutting Mats will protect all table-top surfaces from knife cuts. Its smooth surface permits straight, even cutting and the cut mark heals instantly. Each mat is printed with both 1/2" and 1" grid patterns to aid in measuring and aligning work. Available in translucent white, which allows for cutting and trimming on a light box surface.