Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cell Signaling Technology Whitlow/218 Linker (E3U7Q) Rabbit Monoclonal Antibody, PE Conjugate, 50 Tests
SDP

Catalog No. NC2243799
Encompass

This antibody is conjugated to phycoerythrin (PE) and validated for direct flow cytometric analysis in human cells. It is expected to react with cell surface expressed CARs of varying specificity containing a Whitlow/218 linker within the scFv of the extracellular domain.

  • Validated for flow cytometry (live) applications
  • Conjugated to PE fluorescent dye
  • Recombinant monoclonal antibody produced in rabbit
  • Recognizes exogenously expressed levels of scFv-based CARs
  • Supplied in PBS (pH 7.2), less than 0.1% sodium azide, and 2 mg/mL BSA
  • Storage at 4°C, protect from light

This item is not returnable. View return policy
Catalog No. NC2243799 Supplier Cell Signaling Technology Supplier No. 62405S
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This antibody is conjugated to phycoerythrin (PE) and validated for direct flow cytometric analysis in human cells. It is expected to react with cell surface expressed CARs of varying specificity containing a Whitlow/218 linker within the scFv of the extracellular domain.

  • Validated for flow cytometry (live) applications
  • Conjugated to PE fluorescent dye
  • Recombinant monoclonal antibody produced in rabbit
  • Recognizes exogenously expressed levels of scFv-based CARs
  • Supplied in PBS (pH 7.2), less than 0.1% sodium azide, and 2 mg/mL BSA
  • Storage at 4°C, protect from light

This item is not returnable. View return policy
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.