Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cell Signaling Technology Whitlow/218 Linker Monoclonal Antibody, Rabbit, Unconjugated, 20 μL
SDP

Catalog No. 503133366
Encompass

The Whitlow/218 Linker (F2G3S) Rabbit Monoclonal Antibody is designed to detect exogenously expressed levels of single-chain variable fragment (scFv)-based chimeric antigen receptors (CARs) containing the Whitlow/218 linker. This antibody is produced by immunizing animals with a synthetic peptide corresponding to residues of the Whitlow/218 linker, a sequence frequently utilized to improve resistance to aggregation and proteolysis in scFvs. This product is for research use only.

  • Host species: Rabbit
  • Reactivity: All species expected
  • Sensitivity: Transfected only
  • Applications: Western blotting, immunoprecipitation, immunohistochemistry (paraffin), flow cytometry (fixed/permeabilized), and flow cytometry (live)
  • Storage: 10 mM sodium HEPES (pH 7.5), 150 mM NaCl, 100 μg/mL BSA, 50% glycerol, and <0.02% sodium azide; store at −20°C

Catalog No. 50-313-3366 Supplier Cell Signaling Technology Supplier No. 47414T
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Whitlow/218 Linker (F2G3S) Rabbit Monoclonal Antibody is designed to detect exogenously expressed levels of single-chain variable fragment (scFv)-based chimeric antigen receptors (CARs) containing the Whitlow/218 linker. This antibody is produced by immunizing animals with a synthetic peptide corresponding to residues of the Whitlow/218 linker, a sequence frequently utilized to improve resistance to aggregation and proteolysis in scFvs. This product is for research use only.

  • Host species: Rabbit
  • Reactivity: All species expected
  • Sensitivity: Transfected only
  • Applications: Western blotting, immunoprecipitation, immunohistochemistry (paraffin), flow cytometry (fixed/permeabilized), and flow cytometry (live)
  • Storage: 10 mM sodium HEPES (pH 7.5), 150 mM NaCl, 100 μg/mL BSA, 50% glycerol, and <0.02% sodium azide; store at −20°C

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.