Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wells Lamont™ Whizard™ Defender™ 13 Cut-Resistant Gloves
GSA_VA

Catalog No. 19167807C Shop All Wells Lamont Industry Group Products
Change view
Click to view available options
Size:
Medium
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-167-807C Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-167-807C Supplier Wells Lamont™ Supplier No. 135474
  • $36.25
    $16.65 / Each

    Save $19.60

    54% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
  • $189.00
    $86.65 / Pack of 6 EA

    Save $102.35

    54% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Whizard™ Defender™ 13 Cut-Resistant Gloves with high performance fibers and stainless steel combination offer ANSI Cut Level A5 cut resistance. 13-gauge gloves are ideal for handling sharp surfaces, food preparation, and meat and poultry processing.

Patented combination of high-performance fibers and stainless steel creates superior cut resistant properties, eliminating the high cost of lacerations.

  • Standard cuff
  • Ambidextrous
  • Prewashed and preshrunk
  • 10 gauge: ANSI A6
  • Made in the USA

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

ANSI Abrasion Rating ANSI 3
Certifications/Compliance FDA CFR 21, ASTM F2992-15, ASTM D3884-09
Coating Material Uncoated
Color White, Gray
Cut Resistance Rating ANSI A4
Hand Ambidextrous
Latex Free Yes
Material Fibers, Stainless Steel Fiber
Product Type Cut-resistant Gloves
Size Medium
For Use With (Application) Light duty applications
Glove Cuff Color Gray
Glove Cuff Style Plain
Hem Color Blue
Model 135474
Series Whizard™ Defender™ 13
Yarn Gauge 13
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.