Learn More
MSC Whizard Gloves, Metalguard 1880LP, Cut A7, Medium, 1 Pair
Catalog No. 5000788974
Encompass
These industrial gloves offer advanced cut and abrasion resistance, ideal for demanding applications. Constructed from a blend of stainless steel fiber and leather, they feature an uncoated design in a distinctive green and yellow finish.
  • Knit style for comfortable wear
  • Uncoated for direct tactile feedback
  • ANSI/ISEA Cut Resistance Level A7
  • ANSI/ISEA Abrasion Resistance Level 5
  • Medium size for optimal fit
  • Primary material: stainless steel fiber and leather
  • Gauge of 7 for heavy-duty use
  • Knit cuff style ensures a secure fit
  • ANSI/ISEA Puncture Resistance Level 0
  • Kevlar lining for enhanced protection
  • Grain palm and split back leather construction
  • Suitable for wet and oily environments
  • Heavy weight design for demanding tasks
  • Leather palm provides extra abrasion and puncture resistance
Catalog No. 50-007-88974 Supplier MSC Supplier No. 43765627
These industrial gloves offer advanced cut and abrasion resistance, ideal for demanding applications. Constructed from a blend of stainless steel fiber and leather, they feature an uncoated design in a distinctive green and yellow finish.
  • Knit style for comfortable wear
  • Uncoated for direct tactile feedback
  • ANSI/ISEA Cut Resistance Level A7
  • ANSI/ISEA Abrasion Resistance Level 5
  • Medium size for optimal fit
  • Primary material: stainless steel fiber and leather
  • Gauge of 7 for heavy-duty use
  • Knit cuff style ensures a secure fit
  • ANSI/ISEA Puncture Resistance Level 0
  • Kevlar lining for enhanced protection
  • Grain palm and split back leather construction
  • Suitable for wet and oily environments
  • Heavy weight design for demanding tasks
  • Leather palm provides extra abrasion and puncture resistance