Learn More
Wells Lamont Industrial™ Whizard Ultra Talon Cut-resistant Work Glove
Catalog No. 17827315 Shop All Wells Lamont Industry Group Products
Change view
Click to view available options
Size:
Small
Medium
Large
2X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-827-315 Small
17-827-316 Medium
17-827-317 Large
17-827-319 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 17-827-315 Supplier Wells Lamont Industrial™ Supplier No. 136221
Only null left
Add to Cart
Add to Cart
With its new Hi-Vis cuff area for better visibility and ANSI A8 cut resistance, the Whizard Ultra Talon is an ideal anti-cut work glove for your industrial hand protection and safety needs.
  • ANSI Level 3 for abrasion and Category III for particulates
  • Cut resistant Talon gloves are recognized and favored by the meat and poultry industries for high protection and performance
  • Patented yarn composition combines high strength, high-performance materials with stainless steel, creating high tensile strength, and high cut protection
  • Approved for cleanroom use
  • Made in the USA
Specifications
Certifications/Compliance CFR 21 Approved for Food Contact
Construction Seamless Knit
Coating Material Uncoated
Color Gray
Cut Resistance Rating ANSI A8
Hand Ambidextrous
Latex Free Yes
Material Engineered Yarn
Product Type Cut-resistant Gloves
Size Small
Gender Unisex
Glove Cuff Style Elasticated
Lining Unlined
Puncture Rating ANSI/ISEA 105-16 Level 0
Thumb Style Straight