Learn More
Abcam Whole Blood Polymorphonuclear Cell Isolation Kit, 30 Tests
Catalog No. 502205218
Encompass_Preferred
Whole Blood Polymorphonuclear Cell Isolation Kit (ab241001) is unique in the market because it enables the user to isolate Polymorphonuclear cells (PMNs), assess viability of the obtained cell fractions and also determine purity with three simple, easy-to-use reagents.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-220-5218 Supplier Abcam Supplier No. AB24100130TEST
Whole Blood Polymorphonuclear Cell Isolation Kit (ab241001) is unique in the market because it enables the user to isolate Polymorphonuclear cells (PMNs), assess viability of the obtained cell fractions and also determine purity with three simple, easy-to-use reagents.
The product is subject to the following: Abcam Restricted Use Statement