Learn More
Aviva Systems Biology Corp NSD2 Antibody - N-terminal region (ARP33489_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33489_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against WHSC1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human WHSC1. Alternate names: WHS, NSD2, TRX5, MMSET, REIIBPApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-52907