Learn More
Eurofins Discoverx WHSC1 (NSD2), 50 µg. (EPI073)
Supplier:  Eurofins Discoverx EPI073
Encompass
WHSC1 (NSD2) is purified, N terminal 6xHis tagged recombinant WHSC1 enzyme. This enzyme contains amino acids 825-1208. It retains its binding/enzymatic activity and is intended for use in epigenetic assays. This enzyme is provided as 0.765 mg/mL frozen in 50% glycerol. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-200-5295