Learn More
Consolidated Plastics WHT CANISTER 60OZ RED LID 12PK
SDP
Catalog No. NC0779003
Encompass
60 Oz. Canister - 4 3/4 in.Dia x 6 3/8 in.H - Red Lid High Density Polyethylene Canisters are ideal for most food products as well as chemicals and powders. Lids are constructed from FDa acceptable polyethylene. Lids feature 16 vertical ribs for easy gripping and turning. Non-stripping 120mm triple lead thread ensures proper sealing. Color is white. Canisters comply with FDa regulations. Red lids included.
Catalog No. NC0779003 Supplier Consolidated Plastics Supplier No. 42596
60 Oz. Canister - 4 3/4 in.Dia x 6 3/8 in.H - Red Lid High Density Polyethylene Canisters are ideal for most food products as well as chemicals and powders. Lids are constructed from FDa acceptable polyethylene. Lids feature 16 vertical ribs for easy gripping and turning. Non-stripping 120mm triple lead thread ensures proper sealing. Color is white. Canisters comply with FDa regulations. Red lids included.