Learn More
Electronic Imaging Materials Inc 667 Vial CryoLabel 0.88 inch x 0.56 inch,White, 1 inch core, 1000/roll
SDP
Catalog No. NC0825507
Encompass_Preferred
0.88 inch x 0.56 inch CryoLabel will handle extremely cold storage temperatures found in genetics, virology, microbiology and related fields. Resistant to moisture, smudging, and tearing, plus it holds up to alcohol and other chemicals. Designed for thermal transfer printing and durable UV inkjet printing, allowing you to color-code labels. Scratch and smear resistant when printed with T68 premium resin ribbon.
Catalog No. NC0825507 Supplier Electronic Imaging Materials Inc Supplier No. XU100073667
0.88 inch x 0.56 inch CryoLabel will handle extremely cold storage temperatures found in genetics, virology, microbiology and related fields. Resistant to moisture, smudging, and tearing, plus it holds up to alcohol and other chemicals. Designed for thermal transfer printing and durable UV inkjet printing, allowing you to color-code labels. Scratch and smear resistant when printed with T68 premium resin ribbon.